Learn More
2M Lithium Chloride Electrolyte, Electrode Filling Solution, Paragon Scientific™
A high quality analytical reagent manufactured and certified compliant for use in ASTM D 664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.
Brand: Paragon Scientific LCL-30-10
Additional Details : CAS Number : 64-17-5 Weight : 0.50000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- A Lithium Chloride Electrolyte Analytical Reagent manufactured to ASTM D 664 / IP 177.
- Provided with a certificate which includes an estimation of uncertainty of manufacture.
- Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
- Convenient 30mL bottle size for direct filling into the electrode.
- Multi pack option available
- Cost effective and time saving
- Certificate of Manufacture & MSDS included
- 6 Month shelf life
Specifications
10 x 30 mL |
Safety and Handling
- X10 30ML LCL-30-10 Lithium Chloride Electrolyte Reagent
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : ethanol, lithium chloride