missing translation for 'onlineSavingsMsg'
Learn More

2M Lithium Chloride Electrolyte, Electrode Filling Solution, Paragon Scientific™

A high quality analytical reagent manufactured and certified compliant for use in ASTM D 664 / IP 177, the Standard Test Method for Acid Number of Petroleum products by Potentiometric Titration.

Brand:  Paragon Scientific LCL-30-5


 View more versions of this product

Product Code. 16070200

  • £164.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A Lithium Chloride Electrolyte Analytical Reagent manufactured to ASTM D 664 / IP 177.
  • Provided with a certificate which includes an estimation of uncertainty of manufacture.
  • Ensures full compliance with ASTM and IP test method requirements for reagent preparation.
  • Convenient 30mL bottle size for direct filling into the electrode.
  • Multi pack option available
  • Cost effective and time saving
  • Certificate of Manufacture & MSDS included
  • 6 Month shelf life
Specifications

Specifications

5 x 30 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2M Lithium Chloride Electrolyte, Electrode Filling Solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : Ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers