missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | Amber to Colorless |
Enantiomeric Excess | 97.5% min. (GC) |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 500 mg |
Synonym | 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid methyl ester, 2r,4s-1-tert-butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid, 2r,4s-1-tert-butyl2-methyl4-hydroxypiperidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-cis-4-hydroxy-l-pipecolinic acid methyl ester, 1-o-tert-butyl 2-o-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, o1-tert-butyl o2-methyl 2r,4s-4-oxidanylpiperidine-1,2-dicarboxylate, tert-butyl 4s,2r-4-hydroxy-2-methoxycarbonyl piperidinecarboxylate, 2r-4beta-hydroxypiperidine-1,2beta-dicarboxylic acid 1-tert-butyl 2-methyl ester |
Formula Weight | 259.3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'