missing translation for 'onlineSavingsMsg'
Learn More
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%, Thermo Scientific Chemicals Product Code.: 15418306

(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%, Thermo Scientific Chemicals

Product Code. 15418306
500 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 15418306

Brand: Thermo Scientific Alfa Aesar H63420.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1044256-04-3
Molecular Formula C15H21O2P
Molecular Weight (g/mol) 264.305
MDL Number MFCD09842713
InChI Key JAKXTLMNIHPLEU-VXGBXAGGSA-N
Synonym 2r,5r-1-2-1,3-dioxolan-2-yl phenyl-2,5-dimethylphospholane, r,r-me-rajphos, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-1,3-dioxolane, 2-2-2s,5s-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, 2-2-2r,5r-2,5-dimethyl-1-phospholano phenyl 1,3-dioxolane, min
PubChem CID 71310562
IUPAC Name 2-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-1,3-dioxolane
SMILES CC1CCC(P1C2=CC=CC=C2C3OCCO3)C

Specifications

Color Colorless to Yellow
Density 1.105 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.572
Quantity 500 mg
Sensitivity Air and moisture sensitive
Formula Weight 264.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2R,5R)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.