missing translation for 'onlineSavingsMsg'
Learn More
(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 10083520

(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10083520
100 mg, Glass bottle
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10083520

Brand: Thermo Scientific Acros 290081000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 118758-48-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 234°C to 236°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C11H13NO2
MDL Number MFCD00142984
Quantity 100 mg
Specific Rotation Condition + 84.00 (25.00°C c=0.75,6 N HCl)
Specific Rotation + 84
Synonym 3-phenyl-l-proline, 3-phenyl proline, 2s-3-phenylpyrrolidine-2-carboxylic acid
InChI Key VDEMEKSASUGYHM-QVDQXJPCSA-N
SMILES C1CNC(C1C2=CC=CC=C2)C(=O)O
IUPAC Name (2R)-3-phenylpyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 191.23
PubChem CID 133109024
Formula Weight 191.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2S, 3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0283418 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number290081000
compliance-icons
Product Identifier
  • (2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2S,3R)-3-Phenylpyrrolidine-2-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.