Learn More
(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific Chemicals
£28.26 - £97.80
Chemical Identifiers
CAS | 70987-78-9 |
---|---|
Molecular Formula | C10H12O4S |
Molecular Weight (g/mol) | 228.27 |
MDL Number | MFCD00064489 |
InChI Key | NOQXXYIGRPAZJC-VIFPVBQESA-N |
Synonym | s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s |
PubChem CID | 153296 |
IUPAC Name | [(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
70987-78-9 | |
228.27 | |
NOQXXYIGRPAZJC-VIFPVBQESA-N | |
153296 | |
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 |
C10H12O4S | |
MFCD00064489 | |
s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s | |
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate |
Specifications
70987-78-9 | |
100.0 | |
White | |
Authentic | |
C10H12O4S | |
+ 17.00 (20.00°C c=2.5,CHCl3) | |
s-glycidyl tosylate, 2s-+-glycidyl tosylate, s-+-glycidyl tosylate, +-glycidyl tosylate, s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, 2s-oxiran-2-ylmethyl 4-methylbenzenesulfonate, ccris 6400, s-glycidyltosylate, 2s-glycidyl tosylate, oxiranemethanol, 4-methylbenzenesulfonate, s | |
NOQXXYIGRPAZJC-VIFPVBQESA-N | |
[(2S)-oxiran-2-yl]methyl 4-methylbenzenesulfonate | |
153296 | |
99+% | |
(2S)-(+)-Glycidyl tosylate |
99.0 | |
45°C to 48°C | |
>110°C | |
99% min. (GC) | |
MFCD00064489 | |
+ 17 | |
Solubility in water: decomposes. Other solubilities: soluble in chloroform | |
CC1=CC=C(C=C1)S(=O)(=O)OCC2CO2 | |
228.27 | |
228.27 | |
Fluffy Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause cancer.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medi
GHS Signal Word: Danger
- (2S)-(+)-Glycidyl tosylate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H341-Suspected of causing genetic defects.
- H411-Toxic to aquatic life with long lasting effects.
- H318-Causes serious eye damage.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only