missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1,1,1-Tributylstannyl)pyridine, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 368.15 |
Percent Purity | 90% |
Chemical Name or Material | 3-(1, 1, 1-Tributylstannyl)pyridine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(1,1,1-Tributylstannyl)pyridine, 90%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction