missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-Pyrrolyl)-2-thiophenemethanol, 97%, Thermo Scientific™
£47.30 - £141.00
Chemical Identifiers
CAS | 107073-27-8 |
---|---|
Molecular Formula | C9H9NOS |
Molecular Weight (g/mol) | 179.237 |
MDL Number | MFCD00052578 |
InChI Key | SMSUHWYHSLZKHS-UHFFFAOYSA-N |
Synonym | 3-pyrrol-1-yl thiophene-2-methanol, 3-1h-pyrrol-1-yl-2-thienyl methanol, 3-pyrrol-1-ylthiophen-2-yl methanol, 3-1h-pyrrol-1-yl thiophen-2-yl methanol, 3-pyrrol-1-yl thiophen-2-yl methanol, 3-1h-pyrrol-1-yl thien-2-yl methanol, 3-pyrrolyl-2-thienyl methan-1-ol, 3-1-pyrrolyl-2-thiophenemethanol, 3-1-pyrrolyl-2-thiophenyl methanol |
PubChem CID | 342464 |
IUPAC Name | (3-pyrrol-1-ylthiophen-2-yl)methanol |
SMILES | C1=CN(C=C1)C2=C(SC=C2)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15477177
|
Thermo Scientific Alfa Aesar
H34243.03 |
1 g |
£47.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487177
|
Thermo Scientific Alfa Aesar
H34243.06 |
5 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
107073-27-8 | |
179.237 | |
SMSUHWYHSLZKHS-UHFFFAOYSA-N | |
342464 | |
C1=CN(C=C1)C2=C(SC=C2)CO |
C9H9NOS | |
MFCD00052578 | |
3-pyrrol-1-yl thiophene-2-methanol, 3-1h-pyrrol-1-yl-2-thienyl methanol, 3-pyrrol-1-ylthiophen-2-yl methanol, 3-1h-pyrrol-1-yl thiophen-2-yl methanol, 3-pyrrol-1-yl thiophen-2-yl methanol, 3-1h-pyrrol-1-yl thien-2-yl methanol, 3-pyrrolyl-2-thienyl methan-1-ol, 3-1-pyrrolyl-2-thiophenemethanol, 3-1-pyrrolyl-2-thiophenyl methanol | |
(3-pyrrol-1-ylthiophen-2-yl)methanol |
Specifications
107073-27-8 | |
C9H9NOS | |
Air Sensitive | |
SMSUHWYHSLZKHS-UHFFFAOYSA-N | |
(3-pyrrol-1-ylthiophen-2-yl)methanol | |
342464 | |
97% |
104°C to 108°C (1.5 mmHg) | |
MFCD00052578 | |
3-pyrrol-1-yl thiophene-2-methanol, 3-1h-pyrrol-1-yl-2-thienyl methanol, 3-pyrrol-1-ylthiophen-2-yl methanol, 3-1h-pyrrol-1-yl thiophen-2-yl methanol, 3-pyrrol-1-yl thiophen-2-yl methanol, 3-1h-pyrrol-1-yl thien-2-yl methanol, 3-pyrrolyl-2-thienyl methan-1-ol, 3-1-pyrrolyl-2-thiophenemethanol, 3-1-pyrrolyl-2-thiophenyl methanol | |
C1=CN(C=C1)C2=C(SC=C2)CO | |
179.237 | |
179.24 | |
3-(1-Pyrrolyl)-2-thiophenemethanol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen