missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-1,2,4-triazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 876728-37-9 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Molecular Formula | C10H8N4 |
MDL Number | MFCD08271931 |
Quantity | 1 g |
Synonym | 3-1h-1,2,4-triazol-1-ylmethyl benzonitrile, 3-1,2,4-triazol-1-ylmethyl benzonitrile, 3-1h-1,2,4-triazol-1-yl methyl benzonitrile, 3-1,2,4-triazolylmethyl benzenecarbonitrile, benzonitrile,3-1h-1,2,4-triazol-1-ylmethyl |
InChI Key | ZYQNZNVPCHZDLT-UHFFFAOYSA-N |
SMILES | N#CC1=CC(CN2C=NC=N2)=CC=C1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'