Learn More
[3-(1H-1,2,4-Triazol-1-ylmethyl)phenyl]methanol, ≥97%, Thermo Scientific™
£25.30 - £85.15
Chemical Identifiers
CAS | 871825-54-6 |
---|---|
Molecular Formula | C10H11N3O |
Molecular Weight (g/mol) | 189.22 |
MDL Number | MFCD08271929 |
InChI Key | PZRKSBSNVMSIJI-UHFFFAOYSA-N |
Synonym | 3-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl phenyl methanol, 3-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl benzyl alcohol, 3-1,2,4-triazolylmethyl phenyl methan-1-ol |
PubChem CID | 18525775 |
SMILES | OCC1=CC=CC(CN2C=NC=N2)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11340627
|
Thermo Scientific Maybridge
CC45409CB |
250 mg |
£25.30
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10642865
|
Thermo Scientific Maybridge
CC45409DA |
1 g |
£85.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
871825-54-6 | |
189.22 | |
PZRKSBSNVMSIJI-UHFFFAOYSA-N | |
18525775 |
C10H11N3O | |
MFCD08271929 | |
3-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl phenyl methanol, 3-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl benzyl alcohol, 3-1,2,4-triazolylmethyl phenyl methan-1-ol | |
OCC1=CC=CC(CN2C=NC=N2)=C1 |
Specifications
871825-54-6 | |
100.0 | |
C10H11N3O | |
3-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl phenyl methanol, 3-1,2,4-triazol-1-ylmethyl phenyl methanol, 3-1h-1,2,4-triazol-1-yl methyl benzyl alcohol, 3-1,2,4-triazolylmethyl phenyl methan-1-ol | |
OCC1=CC=CC(CN2C=NC=N2)=C1 | |
189.22 | |
189.22 | |
[3-(1H-1, 2, 4-Triazol-1-ylmethyl)phenyl]methanol |
94.5 | |
95% | |
MFCD08271929 | |
PZRKSBSNVMSIJI-UHFFFAOYSA-N | |
[3-(1,2,4-triazol-1-ylmethyl)phenyl]methanol | |
18525775 | |
95% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.