missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Imidazol-1-yl)propanoic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 140.14 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1H-imidazol-1-yl)propanoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'