missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-Pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Synonym | 3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl |
IUPAC Name | 3-(1H-pyrrol-1-yl)benzoate |
Formula Weight | 187.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1H-Pyrrol-1-yl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'