missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10212832
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 83140-94-7
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD03086142
InChI Key QQXDXYAGEXWXQU-UHFFFAOYSA-N
Synonym 3-1h-pyrrol-1-yl phenyl methanol, 3-pyrrol-1-yl phenyl methanol, 3-1-pyrrolyl benzyl alcohol, 3-pyrrol-1-ylphenyl methanol, 3-pyrrolylphenyl methan-1-ol, 3-pyrrol-1-yl phenylmethanol, 3-1h-pyrrol-1-yl phenylmethanol, benzenemethanol, 3-1h-pyrrol-1-yl
PubChem CID 2776531
IUPAC Name (3-pyrrol-1-ylphenyl)methanol
SMILES C1=CN(C=C1)C2=CC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [3-(1H-Pyrrol-1-yl)phenyl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.