missing translation for 'onlineSavingsMsg'
Learn More

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride, Thermo Scientific™

Product Code. 10693603
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4462-55-9
Molecular Formula C11H6Cl3NO2
Molecular Weight (g/mol) 290.524
InChI Key IZQGELJKDARDMZ-UHFFFAOYSA-N
Synonym 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonyl chloride, dcimc chloride, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carbonyl chloride, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonylchloride, 4-isoxazolecarbonyl chloride, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid chloride, 3-2,6-dichlorophenyl-5-methyl isoxazole-4-carbonyl chloride, pubchem8683, dsstox_cid_31536, dsstox_rid_97421
PubChem CID 78212
IUPAC Name 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 290.53
Percent Purity 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'