missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11392977
Click to view available options
Quantity:
1 g
5 g

3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Aminoethyl)pyridine has been used in the synthesis of urea.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20173-24-4
Molecular Formula C7H11N2
Molecular Weight (g/mol) 123.18
MDL Number MFCD00191604
InChI Key NAHHNSMHYCLMON-UHFFFAOYSA-O
Synonym 3-2-aminoethyl pyridine, 2-pyridin-3-yl ethanamine, 3-aminoethylpyridine, 3-pyridineethanamine, 2-pyridin-3-yl ethan-1-amine, 2-3-pyridyl ethylamine, 3-pyridineethaneamine, 3-pyridine ethanamine, 2-pyridin-3-yl-ethylamine, 2-3-pyridinyl ethanamine
PubChem CID 854051
SMILES [NH3+]CCC1=CC=CN=C1

Specifications

Boiling Point 119°C to 120°C (19 mmHg)
Odor Amine-like
Refractive Index 1.5518
Quantity 1 g
UN Number UN2735
Beilstein 111288
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
IUPAC Name 2-(pyridin-3-yl)ethan-1-aminium
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 3-(2-Aminoethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'