missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11426396
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-29-2
Molecular Formula C8H22N2O2Si
Molecular Weight (g/mol) 206.36
MDL Number MFCD00054776
InChI Key MQWFLKHKWJMCEN-UHFFFAOYSA-N
Synonym n-2-aminoethyl-3-aminopropylmethyldimethoxysilane, 3-2-aminoethylamino propyl-dimethoxymethylsilane, 3-2-aminoethylamino propyldimethoxymethylsilane, 3-2-aminoethylamino propylmethyldimethoxysilane, n-3-dimethoxymethylsilyl propyl ethylenediamine, 1,2-ethanediamine, n-3-dimethoxymethylsilyl propyl, n-3-dimethoxy methyl silyl propyl ethane-1,2-diamine, n-2-aminoethyl-3-aminopropyltrimethoxysilane, 1,2-ethanediamine, n1-3-dimethoxymethylsilyl propyl
PubChem CID 76482
IUPAC Name N'-[3-[dimethoxy(methyl)silyl]propyl]ethane-1,2-diamine
SMILES CO[Si](C)(CCCNCCN)OC

Specifications

Boiling Point 265°C
Quantity 100 g
UN Number UN2735
Beilstein 2074259
Flash Point 90°C (194°F)
Formula Weight 206.36
Percent Purity 97%
Odor Amine-like
Refractive Index 1.449
Sensitivity Air and moisture sensitive
Density 0.968
Chemical Name or Material 3-(2-Aminoethylamino)propylmethyldimethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'