missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Bromoacetyl)benzonitrile, 95%, Thermo Scientific™
£21.95 - £243.00
Chemical Identifiers
CAS | 50916-55-7 |
---|---|
Molecular Formula | C9H6BrNO |
Molecular Weight (g/mol) | 224.06 |
MDL Number | MFCD00833267 |
InChI Key | XWCGNFLHRINYCE-UHFFFAOYSA-N |
Synonym | 3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile |
PubChem CID | 2735867 |
IUPAC Name | 3-(2-bromoacetyl)benzonitrile |
SMILES | BrCC(=O)C1=CC=CC(=C1)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10722513
|
Thermo Scientific Maybridge
SPB07593DA |
1 g | Amber Glass Bottle |
£21.95
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10021984
|
Thermo Scientific Maybridge
SPB07593EB |
25 g | Amber Glass Bottle |
£243.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
50916-55-7 | |
224.06 | |
XWCGNFLHRINYCE-UHFFFAOYSA-N | |
2735867 | |
BrCC(=O)C1=CC=CC(=C1)C#N |
C9H6BrNO | |
MFCD00833267 | |
3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile | |
3-(2-bromoacetyl)benzonitrile |
Specifications
50916-55-7 | |
C9H6BrNO | |
MFCD00833267 | |
XWCGNFLHRINYCE-UHFFFAOYSA-N | |
3-(2-bromoacetyl)benzonitrile | |
2735867 | |
95% |
95% | |
BrCH2C(O)C6H4CN | |
3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile | |
BrCC(=O)C1=CC=CC(=C1)C#N | |
224.06 | |
224.06 | |
3-(2-Bromoacetyl)benzonitrile |