missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-5-methylphenoxy)piperidine, 98%, Thermo Scientific™

Product Code. 11419770
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902837-30-3
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.716
MDL Number MFCD08061103
InChI Key GRLVAJJXUNOVRA-UHFFFAOYSA-N
Synonym 3-2-chloro-5-methylphenoxy piperidine, 3-2-chloro-5-methyl-phenoxy-piperidine, 1-chloro-4-methyl-2-3-piperidyloxy benzene
PubChem CID 24212031
IUPAC Name 3-(2-chloro-5-methylphenoxy)piperidine
SMILES CC1=CC(=C(C=C1)Cl)OC2CCCNC2

Specifications

Quantity 250 mg
Formula Weight 225.72
Percent Purity 98%
Chemical Name or Material 3-(2-Chloro-5-methylphenoxy)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'