missing translation for 'onlineSavingsMsg'
Learn More

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10455712
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3919-74-2
Molecular Formula C11H7ClFNO3
Molecular Weight (g/mol) 255.63
MDL Number MFCD00055659
InChI Key REIPZLFZMOQHJT-UHFFFAOYSA-N
Synonym 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, fcimic acid, 4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid, pubchem8772, acmc-1ckjh, 3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid
PubChem CID 77520
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 255.63
Percent Purity 97%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'