missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-chlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Quantity | 25 g |
Packaging | Amber Glass Bottle |
Assay Percent Range | 97% |
Formula Weight | 237.64 |
Percent Purity | 97% |
Chemical Name or Material | 3-(2-chlorophenyl)-5-methylisoxazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'