missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-3-furylthio)-4-heptanone, 97%, Thermo Scientific™

Product Code. 15443677
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61295-41-8
Molecular Formula C12H18O2S
Molecular Weight (g/mol) 226.33
MDL Number MFCD01723735
InChI Key GFRRQSASJZMMJC-UHFFFAOYNA-N
Synonym 3-2-methyl-3-furylthio-4-heptanone, 3-2-methylfuran-3-yl thio heptan-4-one, fema no. 3570, 3-2-methyl-3-furyl thio-4-heptanone, 1,3-diethylacetonyl 3-methyl-3-furyl sulfide, 3-2-methyl-3-furyl thio heptan-4-one, 4-heptanone, 3-2-methyl-3-furanyl thio, 4-heptanone, 3-2-methyl-3-furyl thio, 3-2-methylfuran-3-yl sulfanylheptan-4-one, 3-2-methyl-3-furanyl thio-4-heptanone
PubChem CID 43598
IUPAC Name 3-(2-methylfuran-3-yl)sulfanylheptan-4-one
SMILES CCCC(=O)C(CC)SC1=C(C)OC=C1

Specifications

Refractive Index 1.5015
Quantity 250 mg
Formula Weight 226.34
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-3-furylthio)-4-heptanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'