missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Morpholinoethoxy)phenyl]methylamine, 95%, Thermo Scientific™

Product Code. 10612045
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857284-08-3
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.315
MDL Number MFCD07772847
InChI Key KRJWFDTWNMICLY-UHFFFAOYSA-N
Synonym 3-2-morpholinoethoxy phenyl methylamine, 3-2-morpholin-4-yl ethoxy phenyl methanamine, 3-2-morpholin-4-yl-ethoxy-benzylamine, 3-2-morpholin-4-yl ethoxy phenyl methylamine, benzenemethanamine, 3-2-4-morpholinyl ethoxy, 1-3-2-morpholin-4-yl ethoxy phenyl methanamine, 3-2-morpholin-4-ylethoxy phenyl methanamine, 3-2-morpholinoethoxy phenylmethylamine, 3-morpholinoethoxy-phenyl methyl amin, 3-2-morpholinoethoxy phenyl methanamine
PubChem CID 7164624
IUPAC Name [3-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES C1COCCN1CCOC2=CC=CC(=C2)CN

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 236.31
Percent Purity 95%
Chemical Name or Material [3-(2-Morpholinoethoxy)phenyl]methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'