missing translation for 'onlineSavingsMsg'
Learn More
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals Product Code.: 10751684

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals

Product Code. 10751684
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10751684

Brand: Thermo Scientific Acros 171010250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264198-47-1, 28048-33-1
Molecular Formula C20H12N4Na2O6S2
Molecular Weight (g/mol) 514.44
MDL Number MFCD00717621
InChI Key DDIHLFCSEHAOJZ-UHFFFAOYSA-L
Synonym 5,6-diphenyl-3-2-pyridyl-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, disodium ferrozine 2-hydrate, 3-2-pyridyl-5,6-bis 4-sulfophenyl-1,2,4-triazine disodium salt hydrateShow MoreShow Less
PubChem CID 91871971
IUPAC Name disodium;4-[3-pyridin-2-yl-6-(4-sulfonatophenyl)-1,2,4-triazin-5-yl]benzenesulfonate;hydrate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 25 g
Solubility Information (5% acetate-buffer pH 5.5) Clear yellow
Formula Weight 514.45
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467293 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number17101015,171010250,171010010,171010050
Lot NumberA0462686 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number17101015,171010250,171010010,171010050
Lot NumberA0458390 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog Number17101015,171010250,171010010,171010050
Lot NumberA0453444 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number17101015,171010250,171010010,171010050
Lot NumberA0392905 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number17101015,171010250,171010010,171010050
compliance-icons
Product Identifier
  • 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid, disodium salt hydrat, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.