Learn More
(3,3,3-Trifluoropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals
£52.46 - £168.84
Chemical Identifiers
CAS | 429-60-7 |
---|---|
Molecular Formula | C6H13F3O3Si |
Molecular Weight (g/mol) | 218.25 |
MDL Number | MFCD00039266 |
InChI Key | JLGNHOJUQFHYEZ-UHFFFAOYSA-N |
Synonym | trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3 |
PubChem CID | 67933 |
IUPAC Name | trimethoxy(3,3,3-trifluoropropyl)silane |
SMILES | CO[Si](CCC(F)(F)F)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349315
|
Thermo Scientific Alfa Aesar
L16671.06 |
5 g |
£52.46
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359315
|
Thermo Scientific Alfa Aesar
L16671.14 |
25 g |
£168.84
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Chemical Identifiers
429-60-7 | |
218.25 | |
JLGNHOJUQFHYEZ-UHFFFAOYSA-N | |
67933 | |
CO[Si](CCC(F)(F)F)(OC)OC |
C6H13F3O3Si | |
MFCD00039266 | |
trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3 | |
trimethoxy(3,3,3-trifluoropropyl)silane |
Specifications
144°C | |
C6H13F3O3Si | |
UN1993 | |
38°C (100°F) | |
JLGNHOJUQFHYEZ-UHFFFAOYSA-N | |
trimethoxy(3,3,3-trifluoropropyl)silane | |
67933 | |
97% | |
Moisture sensitive | |
(3,3,3-Trifluoropropyl)trimethoxysilane |
429-60-7 | |
MFCD00039266 | |
1762962 | |
trimethoxy 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropyl trimethoxysilane, 3,3,3-trifluoropropyltrimethoxysilane, silane, trimethoxy 3,3,3-trifluoropropyl, 1,1,1-trifluoro-3-trimethoxysilyl propane, silane, trimethoxy trifluoropropyl, silane,trimethoxy 3,3,3-trifluoropropyl, acmc-1ajbz, fas-3, cf3 ch2 2si och3 3 | |
CO[Si](CCC(F)(F)F)(OC)OC | |
218.25 | |
218.25 | |
1.355 | |
1.14 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 207-059-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.