missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 219.0°C to 226.0°C |
Color | White to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 97+% |
Packaging | Plastic Bottle |
Quantity | 2.5 kg |
Solubility Information | Solubility in water: reacts. Other solubilities: soluble in dmf (1g/10mL) |
Sublimation Point | 0220.0°C |
Show More |
Safety and Handling
Product Identifier
- 3,3',4,4'-Benzophenonetetracarboxylic dianhydride
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1B
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'