missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity, Thermo Scientific Chemicals

Product Code. 15485795
Click to view available options
Quantity:
125 mL
500 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase-coupled systems, particularly in ELISA techniques, blue result
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Color Colorless to Yellow
Odor Alcohol-like
Quantity 1000 mL
UN Number UN2810
Beilstein 2808541
Formula Weight 240.35
Physical Form Liquid
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine soln., Ready-to-Use, precipitating, high sensitivity

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'