Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 1-Way Vacuum Stopcocks
Brand: Lenz Laborglasinstrumente™ 02901402
Additional Details : Weight : 0.50000kg
Specifications
1-way Vacuum Stopcock | |
Borosilicate Glass | |
Straight side arms and hollow Glass plug |
2.5 mm | |
DIN 12540-1 |