Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
£31.20 - £59.05
Product Code | Brand | Diameter (Metric) | Length (Metric) | Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Packaging | Price | Quantity & Availability | |||
11863380
|
DWK Life Sciences
A610010000911 |
10 mm | 100 mm | 1Each |
£31.20
Pack of 100 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
11862373
|
DWK Life Sciences
A613014000911 |
14 mm | 130 mm | 100Pack |
£42.62
Pack of 100 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
11882373
|
DWK Life Sciences
A613016001211 |
16 mm | 130 mm | 100Pack |
£39.45
Pack of 100 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
11337544
|
DWK Life Sciences
A618018001211 |
18 mm | 180 mm | 100Pack |
£49.35
Pack of 100 |
In Stock
Log in to see stock. |
|||
11802383
|
DWK Life Sciences
A618020001211 |
20 mm | 180 mm | 100Pack |
£51.90
Pack of 100 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
11317544
|
DWK Life Sciences
A615020001211 |
20 mm | 150 mm | 100Pack |
£59.05
Pack of 100 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
11812363
|
DWK Life Sciences
A615025001221 |
25 mm | 150 mm | 50Pack |
£41.25
Pack of 50 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||
11822363
|
DWK Life Sciences
A620025001211 |
25 mm | 200 mm | 50Pack |
£41.05
Pack of 50 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
10676423
|
DWK Life Sciences
A620030001211 |
30 mm | 200 mm | 50Pack |
£48.10
Pack of 50 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
Description
- Disposable
- Borosilicate glass
- Excellent resistance to temperature changes
- Autoclavable
Specifications
Round bottom | |
Test Tube |