Learn More
BRAND™ BLAUBRAND™ Class A Volumetric Flask with Three Marks
The volumetric flask with three marks is used to check the functioning of a dispenser. The mark in the middle shows the nominal volume, the upper and lower marks show the error limits as specified in the table below. If the error limit is exceeded even with repeated measurements, the dispenser is out of specification.
Brand:  BRAND™ 38204
 View more versions of this product
Product Code. 10319073
  • £131.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Borosilicate Glass.
  • Beaded rim prevents slippage when suspended by a clamp.
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Batch number and individual certificate included.
  • Dakks calibration certificate available upon request.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN EN ISO 1042. Calibrated to contain (TC, In). Includes DAkkS-certificate
10 mL
NS 10/19
Upper/lower mark: 0.07 mL
Borosilicate Glass
A