missing translation for 'onlineSavingsMsg'
Learn More
3,3'-Di-n-pentyloxadicarbocyanine iodide, 96%, Thermo Scientific™ Product Code.: 15466987

3,3'-Di-n-pentyloxadicarbocyanine iodide, 96%, Thermo Scientific™

Product Code. 15466987
250 mg
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15466987

Brand: Thermo Scientific Alfa Aesar H34077.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53213-92-6
Molecular Formula C29H35IN2O2
Molecular Weight (g/mol) 570.515
MDL Number MFCD07433847
InChI Key WSRFDTBIGBFQOK-UHFFFAOYSA-M
Synonym 3-pentyl-2-1e,3e-5-3-pentyl-1,3-benzoxazol-2 3h-ylidene-1,3-pentadienyl-1,3-benzoxazol-3-ium iodide, 3-pentyl-2-5-3-pentyl-1,3-benzoxazol-3-ium-2-yl penta-2,4, 3-pentyl-2-1e,3e,5z-5-3-pentylbenzo d oxazol-2 3h-ylidene penta-1,3-dien-1-yl benzo d oxazol-3-ium iodide, 3-pentyl-2-5-3-pentyl-1,3-benzoxazol-2 3h-ylidene penta-1,3-dien-1-yl-1,3-benzoxazol-3-ium iodide
PubChem CID 53425759
IUPAC Name 3-pentyl-2-[5-(3-pentyl-1,3-benzoxazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzoxazole;iodide
SMILES CCCCCN1C2=CC=CC=C2OC1=CC=CC=CC3=[N+](C4=CC=CC=C4O3)CCCCC.[I-]

Specifications

Melting Point ∽187°C (decomposition)
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 570.52
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-pentyloxadicarbocyanine iodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,3'-Di-n-pentyloxadicarbocyanine iodide, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.