missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Diaminobenzidine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3,3'-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.
Solubility
Soluble in water (partly).
Notes
Light sensitive. Incompatible with oxidizing agents and heat.
3,3′-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.
Solubility
Soluble in water (partly).
Notes
Light sensitive. Incompatible with oxidizing agents and heat.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 175°C to 177°C |
Quantity | 5 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water (partly). |
Formula Weight | 214.27 |
Percent Purity | 99% |
Chemical Name or Material | 3,3'-Diaminobenzidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'