missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water
Brand: Thermo Scientific Chemicals 112090010
View more versions of this product
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 3'-Diaminobenzidine tetrahydrochloride hydrate | |
96.0 | |
300.0°C | |
10% max. (120°C) | |
96% min. (on dry substance) (Argentometry) | |
C12H18Cl4N4 | |
MFCD08273058 | |
13, 340 | |
Solubility in water: moderately soluble. Other solubilities: insoluble in most common organic solvents | |
Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1 | |
360.10 | |
360.11 | |
Crystalline Powder |
868272-85-9 | |
100.0 | |
Brown-Purple to Gray | |
Authentic | |
Glass bottle | |
4HCl·xH2O | |
1 g | |
3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se | |
KJDSORYAHBAGPP-UHFFFAOYSA-N | |
4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate | |
23191111 | |
97% |
RUO – Research Use Only