Learn More
Thermo Scientific Chemicals 3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water
£24.90 - £178.00
Specifications
Chemical Name or Material | 3, 3'-Diaminobenzidine tetrahydrochloride hydrate |
---|---|
CAS | 868272-85-9 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 300.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10532364
|
Thermo Scientific Chemicals
112090010 |
1 g | Glass bottle |
£24.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10283990
|
Thermo Scientific Chemicals
112090050 |
5 g | Glass bottle |
£53.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10010810
|
Thermo Scientific Chemicals
112090250 |
25 g | Glass bottle |
£178.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 3'-Diaminobenzidine tetrahydrochloride hydrate | |
96.0 | |
300.0°C | |
10% max. (120°C) | |
96% min. (on dry substance) (Argentometry) | |
C12H18Cl4N4 | |
MFCD08273058 | |
3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se | |
KJDSORYAHBAGPP-UHFFFAOYSA-N | |
4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate | |
23191111 | |
97% |
868272-85-9 | |
100.0 | |
Brown-Purple to Gray | |
Authentic | |
Glass bottle | |
4HCl·xH2O | |
13, 340 | |
Solubility in water: moderately soluble. Other solubilities: insoluble in most common organic solvents | |
Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1 | |
360.10 | |
360.11 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-018-9
missing translation for 'rtecsNumber' : DV8753000
missing translation for 'tsca' : TSCA
RUO – Research Use Only