Learn More
3,3'-Diethylthiadicarbocyanine iodide, Thermo Scientific Chemicals
As a solvent soluble laser dye
£39.60 - £406.00
Chemical Identifiers
CAS | 514-73-8 |
---|---|
Molecular Formula | C23H24IN2S2+ |
Molecular Weight (g/mol) | 519.483 |
MDL Number | MFCD00074829 |
InChI Key | MNQDKWZEUULFPX-UHFFFAOYSA-N |
Synonym | 3,3'-diethylthiadicarbocyanine iodide |
PubChem CID | 91869351 |
IUPAC Name | (2E)-3-ethyl-2-[(2E,4E)-5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;hydroiodide |
SMILES | CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360518
|
Thermo Scientific Alfa Aesar
043851.02 |
0.5 g |
£39.60
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370518
|
Thermo Scientific Alfa Aesar
043851.04 |
2 g |
£114.00
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11350518
|
Thermo Scientific Alfa Aesar
043851.09 |
10 g |
£406.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a solvent soluble laser dye. DTDCi is used as fluorescent contrast agent in three-dimensional fluorescence lifetime tomography and as a dye for rapid genetic screening. It is a potential sensitive probe useful for fluorometric analysis of transferable membrane pores and effects of membrane potential on Na cotransports in eel intestinal brush-border membrane vesicles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
514-73-8 | |
519.483 | |
MNQDKWZEUULFPX-UHFFFAOYSA-N | |
91869351 | |
CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I |
C23H24IN2S2+ | |
MFCD00074829 | |
3,3'-diethylthiadicarbocyanine iodide | |
(2E)-3-ethyl-2-[(2E,4E)-5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;hydroiodide |
Specifications
514-73-8 | |
Green | |
MFCD00074829 | |
3838938 | |
3,3'-diethylthiadicarbocyanine iodide | |
MNQDKWZEUULFPX-UHFFFAOYSA-N | |
(2E)-3-ethyl-2-[(2E,4E)-5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;hydroiodide | |
91869351 | |
99% | |
3,3'-Diethylthiadicarbocyanine iodide |
248°C (decomposition) | |
C23H24IN2S2+ | |
UN2811 | |
14,3369 | |
Soluble in methanol | |
CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I | |
519.483 | |
518.47 | |
Powder |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 208-186-7
missing translation for 'rtecsNumber' : DL7060000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only