missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylthiatricarbocyanine perchlorate, 96%, Thermo Scientific Chemicals

Product Code. 15448275
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22268-66-2
Melting Point 180°C to 182°C
Molecular Formula C25H25ClN2O4S2
MDL Number MFCD00041995
Quantity 500 mg
Synonym DTTC perchlorate
InChI Key PZQHZVAROUTGBU-UHFFFAOYSA-M
SMILES CCN1C2=CC=CC=C2SC1=CC=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.[O-]Cl(=O)(=O)=O
IUPAC Name (2E)-3-ethyl-2-[(2E,4E,6E)-7-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)hepta-2,4,6-trienylidene]-1,3-benzothiazole;perchlorate
Molecular Weight (g/mol) 517.055
PubChem CID 9914540
Formula Weight 517.07
Percent Purity 96%
Chemical Name or Material 3,3'-Diethylthiatricarbocyanine perchlorate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.