missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals

£55.38 - £193.97

Chemical Identifiers

CAS 917-92-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.13
MDL Number MFCD00008852
InChI Key PPWNCLVNXGCGAF-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
PubChem CID 13512
IUPAC Name 3,3-dimethylbut-1-yne
SMILES CC(C)(C)C#C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10144850
View Documents Promotion Details
Thermo Scientific Acros
223440050
5 mL Glass bottle
£55.38
5mL
Please to purchase this item. Need a web account? Register with us today!
10341274
View Documents
Thermo Scientific Acros
223440250
25 mL Glass bottle
£193.97
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

917-92-0
82.13
PPWNCLVNXGCGAF-UHFFFAOYSA-N
13512
CC(C)(C)C#C
C6H10
MFCD00008852
3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
3,3-dimethylbut-1-yne

Specifications

917-92-0
100.0
Colorless
33.0°C to 38.0°C
Authentic
C6H10
1.3730 to 1.3750
01, 256
3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
PPWNCLVNXGCGAF-UHFFFAOYSA-N
3,3-dimethylbut-1-yne
13512
98%
3, 3-Dimethyl-1-butyne
97.5
-78.0°C
0.6600g/mL
−034°C
97.5% min. (GC)
(CH3)3CC≡CH
MFCD00008852
0.66
Solubility in water: insoluble. Other solubilities: soluble in chloroform,benzene,toluene
CC(C)(C)C#C
82.13
82.13
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautio

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 213-035-3

Product Identifier
  • 3,3-Dimethyl-1-butyne

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only