missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3,3-Dimethyl-2-butylamine, 98%, Thermo Scientific Chemicals

Product Code. 11474533
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(±)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3850-30-4
Molecular Formula C6H16N
Molecular Weight (g/mol) 102.20
MDL Number MFCD00008078
InChI Key DXSUORGKJZADET-YFKPBYRVSA-O
Synonym 1,2,2-trimethylpropylamine, 2-amino-3,3-dimethylbutane, 2-butanamine, 3,3-dimethyl, 3,3-dimethyl-2-aminobutane, 3,3-dimethyl-2-butanamine, 3-amino-2,2-dimethylbutane, propylamine, 1,2,2-trimethyl, 3,3-dimethyl-2-butylamine, 3,3-dimethylbut-2-ylamine, 3,3-dimethyl-2-butyl amine
PubChem CID 520907
IUPAC Name 3,3-dimethylbutan-2-amine
SMILES C[C@H]([NH3+])C(C)(C)C

Specifications

Quantity 1 g
UN Number UN2733
Formula Weight 101.19
Percent Purity 98%
Chemical Name or Material (±)-3,3-Dimethyl-2-butylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'