missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylacryloyl chloride, 97%

Product Code. p-115506 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10387880 5 g Glass bottle 5g
10153860 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387880 Supplier Thermo Scientific Acros Supplier No. 179730050
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3350-78-5 | C5H7ClO | 118.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3350-78-5
Molecular Formula C5H7ClO
Molecular Weight (g/mol) 118.56
MDL Number MFCD00000730
InChI Key BDUBTLFQHNYXPC-UHFFFAOYSA-N
Synonym 3,3-dimethylacryloyl chloride, 3-methylcrotonoyl chloride, 3-methyl-but-2-enoyl chloride, 3,3-dimethylacrylyl chloride, 2-butenoyl chloride, 3-methyl, 3-methyl-2-butenoyl chloride, 3,3-dimethylacryloylchloride, dimethylacrylyl chloride, dimethyl acryloyl chloride, acmc-1cpb0
PubChem CID 102394
IUPAC Name 3-methylbut-2-enoyl chloride
SMILES CC(C)=CC(Cl)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 145.0°C to 147.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCOCl
Refractive Index 1.4750 to 1.4780
Quantity 5 g
Beilstein 02, 433
Specific Gravity 1.06
Solubility Information Solubility in water: reacts
Formula Weight 118.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylacryloyl chloride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethylacryloyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.