missing translation for 'onlineSavingsMsg'
Learn More

(3,3-Dimethylallyl)triphenylphosphonium bromide, 98+%, Thermo Scientific™

Product Code. 11300084
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1530-34-3
Molecular Formula C23H24BrP
Molecular Weight (g/mol) 411.32
MDL Number MFCD00051864
InChI Key FTMCNCGWNJMMQS-UHFFFAOYSA-M
Synonym 3,3-dimethylallyl triphenylphosphonium bromide, 3-methylbut-2-enyl triphenylphosphonium bromide, 3-methyl-2-butenyl triphenyl-phosphonium bromide, acmc-20apor, 3,3-dimethylallyl-triphenylphosphonium bromide, 3,3-dimethylallyl-triphenyl-phosphonium bromide, 3-methylbut-2-enyl triphenyl phosphanium bromide, 3-methylbut-2-en-1-yl triphenylphosphanium bromide, phosphonium, 3-methyl-2-buten-1-yl triphenyl-, bromide 1:1
PubChem CID 3083722
IUPAC Name 3-methylbut-2-enyl(triphenyl)phosphanium;bromide
SMILES [Br-].CC(C)=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 235°C to 239°C
Quantity 5 g
Beilstein 3641199
Sensitivity Hygroscopic
Formula Weight 411.33
Percent Purity ≥98%
Chemical Name or Material (3,3-Dimethylallyl)triphenylphosphonium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.