missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylbutylamine, 95%

Product Code. p-3593270 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10005863

Brand: Thermo Scientific Acros 179380010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15673-00-4 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15673-00-4
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008201
InChI Key GPWHFPWZAPOYNO-UHFFFAOYSA-N
PubChem CID 19709
IUPAC Name 3,3-dimethylbutan-1-amine
SMILES CC(C)(C)CCN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7520g/mL
Boiling Point 114°C to 116°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCH2CH2NH2
Refractive Index 1.4120 to 1.4160 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.752
Synonym 3,3-dimethylbutylamine, 1-butanamine, 3,3-dimethyl, 3,3-dimethyl butylamine, 3,3-dimethyl-butylamine, 3,3-dimethylbutyamine, 3,3 dimethylbutylamine, acmc-1btoo, 3,3-dimethylbutyl amine, 1-butanamine,3,3-dimethyl
Formula Weight 101.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethylbutylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethylbutylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.