Learn More
3,3-Dimethylglutaric acid, 98%, Thermo Scientific Chemicals
£34.67 - £452.25
Chemical Identifiers
CAS | 4839-46-7 |
---|---|
Molecular Formula | C7H12O4 |
Molecular Weight (g/mol) | 160.17 |
MDL Number | MFCD00002716 |
InChI Key | DUHQIGLHYXLKAE-UHFFFAOYSA-N |
Synonym | 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid |
PubChem CID | 20984 |
ChEBI | CHEBI:68502 |
IUPAC Name | 3,3-dimethylpentanedioic acid |
SMILES | CC(C)(CC(O)=O)CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10467490
|
Thermo Scientific Acros
116240250 |
25 g | Glass bottle |
£34.67
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10743651
|
Thermo Scientific Acros
116241000 |
100 g | Glass bottle |
£102.51
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321514
|
Thermo Scientific Acros
116245000 |
500 g | Glass bottle |
£452.25
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4839-46-7 | |
160.17 | |
DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
20984 | |
3,3-dimethylpentanedioic acid |
C7H12O4 | |
MFCD00002716 | |
3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
CHEBI:68502 | |
CC(C)(CC(O)=O)CC(O)=O |
Specifications
4839-46-7 | |
100.0 | |
White to Beige | |
Authentic | |
C7H12O4 | |
MFCD00002716 | |
3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
3,3-dimethylpentanedioic acid | |
20984 | |
160.17 | |
Fine Crystalline Powder |
97.5 | |
98.0°C to 102.0°C | |
290.0°C | |
98% | |
HO2CCH2C(CH3)2CH2CO2H | |
02, 684 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether,sparingly soluble in benzene and chloroform,insoluble in low-boiling petroleum fractions | |
CC(C)(CC(O)=O)CC(O)=O | |
160.17 | |
CHEBI:68502 | |
98% | |
3, 3-Dimethylglutaric acid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 225-425-
- 3,3-Dimethylglutaric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.