missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylpentane, 99%, Thermo Scientific Chemicals

£42.49 - £107.40

Chemical Identifiers

CAS 562-49-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.205
MDL Number MFCD00009322
InChI Key AEXMKKGTQYQZCS-UHFFFAOYSA-N
Synonym pentane, 3,3-dimethyl, pentane,3,3-dimethyl, unii-r1rh0vbd2l, r1rh0vbd2l, 3,3,-dimethylpentane, acmc-1b14l, dsstox_cid_29275, dsstox_rid_83394, dsstox_gsid_49319, 3,3-dimethylpentane
PubChem CID 11229
IUPAC Name 3,3-dimethylpentane
SMILES CCC(C)(C)CC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11320308
View Documents
Thermo Scientific Alfa Aesar
043706.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11310308
View Documents
Thermo Scientific Alfa Aesar
043706.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

3,3-Dimethylpentane Is used in organic synthesis & as gasoline component, as Pharmaceutical intermediates. It was used in the development and evaluation of the epiocular-eye irritation test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

562-49-2
100.205
AEXMKKGTQYQZCS-UHFFFAOYSA-N
11229
CCC(C)(C)CC
C7H16
MFCD00009322
pentane, 3,3-dimethyl, pentane,3,3-dimethyl, unii-r1rh0vbd2l, r1rh0vbd2l, 3,3,-dimethylpentane, acmc-1b14l, dsstox_cid_29275, dsstox_rid_83394, dsstox_gsid_49319, 3,3-dimethylpentane
3,3-dimethylpentane

Specifications

562-49-2
0.69g/mL
−6°C (21°F)
MFCD00009322
pentane, 3,3-dimethyl, pentane,3,3-dimethyl, unii-r1rh0vbd2l, r1rh0vbd2l, 3,3,-dimethylpentane, acmc-1b14l, dsstox_cid_29275, dsstox_rid_83394, dsstox_gsid_49319, 3,3-dimethylpentane
CCC(C)(C)CC
100.205
100.21
Liquid
-135°C
86°C
C7H16
UN1206
AEXMKKGTQYQZCS-UHFFFAOYSA-N
3,3-dimethylpentane
11229
99%
3,3-Dimethylpentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only