missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Hydroxyphenyl)propionic acid, 98+%

Product Code. 11394524
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11394524

Brand: Thermo Scientific Alfa Aesar L01279.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 621-54-5 | C9H10O3 | 166.176 g/mol

3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

Solubility
Soluble in chloroform and methanol.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-54-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016554
InChI Key QVWAEZJXDYOKEH-UHFFFAOYSA-N
Synonym 3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl propionic acid, dihydro-3-coumaric acid, benzenepropanoic acid, 3-hydroxy, 3-m-hydroxyphenyl propionic acid, 3-hydroxybenzenepropanoic acid, 3-hydroxyphenylpropionic acid, m-hydroxyphenylpropionic acid, beta-m-hydroxyphenyl propionic acid, hydrocinnamic acid, m-hydroxy
PubChem CID 91
ChEBI CHEBI:1427
IUPAC Name 3-(3-hydroxyphenyl)propanoic acid
SMILES C1=CC(=CC(=C1)O)CCC(=O)O

Specifications

Melting Point 110°C to 114°C
Quantity 5 g
Beilstein 1947445
Solubility Information Soluble in chloroform and methanol.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-(3-Hydroxyphenyl)propionic acid
compliance-icons
Product Identifier
  • 3-(3-Hydroxyphenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-(m-Hydroxyphenyl)propionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.