missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methylphenyl)-3-oxopropanenitrile, 97%, Thermo Scientific™

£18.40 - £54.00

Chemical Identifiers

CAS 53882-81-8
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00067922
InChI Key IVLKDYOTZMFMLO-UHFFFAOYSA-N
Synonym 3-methylbenzoylacetonitrile, 3-3-methylphenyl-3-oxopropanenitrile, 3-toluoylacetonitrile, m-toluoylacetonitrile, 3-oxo-3-m-tolyl propanenitrile, 3-oxo-3-m-tolylpropanenitrile, pubchem12073, maybridge1_004502, acmc-1alb6, 3-m-tolyl-3-oxo-propanenitrile
PubChem CID 143105
IUPAC Name 3-(3-methylphenyl)-3-oxopropanenitrile
SMILES CC1=CC=CC(=C1)C(=O)CC#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10232452
View Documents
Thermo Scientific Maybridge
KM01815DA
1 g Amber Glass Bottle
£18.40
1g
Please to purchase this item. Need a web account? Register with us today!
10292492
View Documents
Thermo Scientific Maybridge
KM01815EA
10 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

53882-81-8
159.188
IVLKDYOTZMFMLO-UHFFFAOYSA-N
143105
CC1=CC=CC(=C1)C(=O)CC#N
C10H9NO
MFCD00067922
3-methylbenzoylacetonitrile, 3-3-methylphenyl-3-oxopropanenitrile, 3-toluoylacetonitrile, m-toluoylacetonitrile, 3-oxo-3-m-tolyl propanenitrile, 3-oxo-3-m-tolylpropanenitrile, pubchem12073, maybridge1_004502, acmc-1alb6, 3-m-tolyl-3-oxo-propanenitrile
3-(3-methylphenyl)-3-oxopropanenitrile

Specifications

53882-81-8
97%
CH3C6H4C(O)CH2CN
3-methylbenzoylacetonitrile, 3-3-methylphenyl-3-oxopropanenitrile, 3-toluoylacetonitrile, m-toluoylacetonitrile, 3-oxo-3-m-tolyl propanenitrile, 3-oxo-3-m-tolylpropanenitrile, pubchem12073, maybridge1_004502, acmc-1alb6, 3-m-tolyl-3-oxo-propanenitrile
CC1=CC=CC(=C1)C(=O)CC#N
159.188
159.19
3-(3-Methylphenyl)-3-oxopropanenitrile
97.0
C10H9NO
MFCD00067922
IVLKDYOTZMFMLO-UHFFFAOYSA-N
3-(3-methylphenyl)-3-oxopropanenitrile
143105
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications