Learn More
3-(3-Piperidinyloxy)benzonitrile, 99%, Thermo Scientific Chemicals
£69.60 - £256.00
Chemical Identifiers
CAS | 902836-93-5 |
---|---|
Molecular Formula | C12H14N2O |
Molecular Weight (g/mol) | 202.257 |
MDL Number | MFCD08061079 |
InChI Key | OSURJDMYHKQISX-UHFFFAOYSA-N |
Synonym | 3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile |
PubChem CID | 22309128 |
IUPAC Name | 3-piperidin-3-yloxybenzonitrile |
SMILES | C1CC(CNC1)OC2=CC=CC(=C2)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419680
|
Thermo Scientific Alfa Aesar
H50939.MD |
250 mg | |||||||
11429680
|
Thermo Scientific Alfa Aesar
H50939.03 |
1 g |
£256.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
902836-93-5 | |
202.257 | |
OSURJDMYHKQISX-UHFFFAOYSA-N | |
22309128 | |
C1CC(CNC1)OC2=CC=CC(=C2)C#N |
C12H14N2O | |
MFCD08061079 | |
3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile | |
3-piperidin-3-yloxybenzonitrile |
Specifications
902836-93-5 | |
MFCD08061079 | |
OSURJDMYHKQISX-UHFFFAOYSA-N | |
3-piperidin-3-yloxybenzonitrile | |
22309128 | |
99% |
C12H14N2O | |
3-3-piperidinyloxy benzonitrile, 3-piperidin-3-yloxy benzonitrile, 3-piperidin-3-yl-oxy benzonitrile, 3-3-piperidyloxy benzenecarbonitrile, 3-piperidin-3-yl oxy benzonitrile | |
C1CC(CNC1)OC2=CC=CC(=C2)C#N | |
202.257 | |
202.26 | |
3-(3-Piperidinyloxy)benzonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only