Learn More
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific Chemicals
£63.10 - £617.30
Chemical Identifiers
CAS | 329214-79-1 |
---|---|
Molecular Formula | C11H16BNO2 |
Molecular Weight (g/mol) | 205.06 |
MDL Number | MFCD03084758 |
InChI Key | XEMDFESAXKSEGI-UHFFFAOYSA-N |
Synonym | 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-pyridineboronic acid pinacol ester, pyridine-3-boronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-3-boronic acid, pinacol ester, 2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine |
PubChem CID | 2734656 |
IUPAC Name | 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10764943
|
Thermo Scientific Acros
367600010 |
1 g | Glass bottle |
£63.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10212812
|
Thermo Scientific Acros
367600050 |
5 g | Glass bottle |
£210.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358843
|
Thermo Scientific Acros
367600250 |
25 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
329214-79-1 | |
205.06 | |
XEMDFESAXKSEGI-UHFFFAOYSA-N | |
2734656 | |
CC1(C)OB(OC1(C)C)C1=CC=CN=C1 |
C11H16BNO2 | |
MFCD03084758 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-pyridineboronic acid pinacol ester, pyridine-3-boronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-3-boronic acid, pinacol ester, 2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
329214-79-1 | |
100.0 | |
White | |
96% min. (GC) | |
MFCD03084758 | |
Solubility in water: insoluble | |
CC1(C)OB(OC1(C)C)C1=CC=CN=C1 | |
205.06 | |
205.06 | |
Crystalline Powder |
96.0 | |
103°C to 108°C | |
Authentic | |
C11H16BNO2 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-pyridineboronic acid pinacol ester, pyridine-3-boronic acid pinacol ester, 3-pyridylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-3-boronic acid, pinacol ester, 2-3-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine | |
XEMDFESAXKSEGI-UHFFFAOYSA-N | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
2734656 | |
97% | |
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
RUO – Research Use Only