Learn More
3-(4,5-Difluoro-2-methoxyphenyl)propionic acid, 97%, Thermo Scientific™
£41.20 - £123.00
Chemische Identifikatoren
CAS | 1092460-69-9 |
---|---|
Molecular Formula | C10H10F2O3 |
Molecular Weight (g/mol) | 216.184 |
MDL Number | MFCD09999478 |
InChI Key | WDTSALDVVUTTSK-UHFFFAOYSA-N |
Synonym | 3-4,5-difluoro-2-methoxyphenyl propanoic acid, 3-4,5-difluoro-2-methoxyphenyl propionic acid, 3-4,5-difluoro-2-methoxyphenyl propionic acid, jrdShow More |
PubChem CID | 57363163 |
IUPAC Name | 3-(4,5-difluoro-2-methoxyphenyl)propanoic acid |
SMILES | COC1=CC(=C(C=C1CCC(=O)O)F)F |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | ||||||
15488075
|
Thermo Scientific Alfa Aesar
H32399.MD |
250 mg | N/A | N/A | N/A | |||||
15478075
|
Thermo Scientific Alfa Aesar
H32399.03 |
1 g | N/A | N/A | N/A | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
1092460-69-9 | |
216.184 | |
WDTSALDVVUTTSK-UHFFFAOYSA-N | |
57363163 | |
COC1=CC(=C(C=C1CCC(=O)O)F)F |
C10H10F2O3 | |
MFCD09999478 | |
3-4,5-difluoro-2-methoxyphenyl propanoic acid, 3-4,5-difluoro-2-methoxyphenyl propionic acid, 3-4,5-difluoro-2-methoxyphenyl propionic acid, jrdShow More | |
3-(4,5-difluoro-2-methoxyphenyl)propanoic acid |
Spezifikation
1092460-69-9 | |
C10H10F2O3 | |
3-4,5-difluoro-2-methoxyphenyl propanoic acid, 3-4,5-difluoro-2-methoxyphenyl propionic acid, 3-4,5-difluoro-2-methoxyphenyl propionic acid, jrd | |
COC1=CC(=C(C=C1CCC(=O)O)F)F | |
216.184 | |
216.18 | |
3-(4,5-Difluoro-2-methoxyphenyl)propionic acid |
90°C to 93°C | |
MFCD09999478 | |
WDTSALDVVUTTSK-UHFFFAOYSA-N | |
3-(4,5-difluoro-2-methoxyphenyl)propanoic acid | |
57363163 | |
97% |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
Recommended Storage : Ambient temperatures
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.