Learn More
3,4,5-Tribromo-1H-pyrazole, 97%, Thermo Scientific Chemicals
£28.50 - £332.00
Chemical Identifiers
CAS | 17635-44-8 |
---|---|
Molecular Formula | C3HBr3N2 |
Molecular Weight (g/mol) | 304.767 |
MDL Number | MFCD00040248 |
InChI Key | TXQKCKQJBGFUBF-UHFFFAOYSA-N |
Synonym | 3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g |
PubChem CID | 627674 |
IUPAC Name | 3,4,5-tribromo-1H-pyrazole |
SMILES | C1(=C(NN=C1Br)Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11409902
|
Thermo Scientific Alfa Aesar
A12031.06 |
5 g |
£28.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419902
|
Thermo Scientific Alfa Aesar
A12031.14 |
25 g |
£113.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429902
|
Thermo Scientific Alfa Aesar
A12031.22 |
100 g | |||||||
Description
2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water. Soluble in Chloroform.
Notes
Store in cool place. Keep away from oxidizing agents.
Chemical Identifiers
17635-44-8 | |
304.767 | |
TXQKCKQJBGFUBF-UHFFFAOYSA-N | |
627674 | |
C1(=C(NN=C1Br)Br)Br |
C3HBr3N2 | |
MFCD00040248 | |
3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g | |
3,4,5-tribromo-1H-pyrazole |
Specifications
17635-44-8 | |
Odorless | |
MFCD00040248 | |
3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g | |
TXQKCKQJBGFUBF-UHFFFAOYSA-N | |
3,4,5-tribromo-1H-pyrazole | |
627674 | |
97% |
184°C to 188°C | |
C3HBr3N2 | |
113501 | |
Insoluble in water. Soluble in Chloroform. | |
C1(=C(NN=C1Br)Br)Br | |
304.767 | |
304.78 | |
3,4,5-Tribromo-1H-pyrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only