Learn More
3,4,5-Trifluorophenol, 97%, Thermo Scientific™
£36.47 - £150.75
Chemical Identifiers
CAS | 99627-05-1 |
---|---|
Molecular Formula | C6H3F3O |
Molecular Weight (g/mol) | 148.084 |
MDL Number | MFCD00042427 |
InChI Key | ZRTWIJKGTUGZJY-UHFFFAOYSA-N |
Synonym | phenol, 3,4,5-trifluoro, 3,4,5-trifluoro-phenol, pubchem2316, 3,4,5-trifluoropheno, acmc-209sdi, ksc486m6r, 3 , 4 , 5-trifluorophenol, phenol, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenol |
PubChem CID | 2777943 |
IUPAC Name | 3,4,5-trifluorophenol |
SMILES | C1=C(C=C(C(=C1F)F)F)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11400148
|
Thermo Scientific Alfa Aesar
B23718.03 |
1 g | N/A | ||||||
11410148
|
Thermo Scientific Alfa Aesar
B23718.06 |
5 g |
£150.75
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
99627-05-1 | |
148.084 | |
ZRTWIJKGTUGZJY-UHFFFAOYSA-N | |
2777943 | |
C1=C(C=C(C(=C1F)F)F)O |
C6H3F3O | |
MFCD00042427 | |
phenol, 3,4,5-trifluoro, 3,4,5-trifluoro-phenol, pubchem2316, 3,4,5-trifluoropheno, acmc-209sdi, ksc486m6r, 3 , 4 , 5-trifluorophenol, phenol, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenol | |
3,4,5-trifluorophenol |
Specifications
99627-05-1 | |
177°C | |
MFCD00042427 | |
8830884 | |
ZRTWIJKGTUGZJY-UHFFFAOYSA-N | |
3,4,5-trifluorophenol | |
2777943 | |
97% |
52°C to 56°C | |
C6H3F3O | |
UN3261 | |
phenol, 3,4,5-trifluoro, 3,4,5-trifluoro-phenol, pubchem2316, 3,4,5-trifluoropheno, acmc-209sdi, ksc486m6r, 3 , 4 , 5-trifluorophenol, phenol, 3,4,5-trifluoro-9ci, 3,4,5-trifluorophenol | |
C1=C(C=C(C(=C1F)F)F)O | |
148.084 | |
148.09 | |
3,4,5-Trifluorophenol |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 436-160-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures