Learn More
3,4,5-Trimethoxyaniline, 97%, Thermo Scientific Chemicals
£30.90 - £169.57
Chemical Identifiers
CAS | 24313-88-0 |
---|---|
Molecular Formula | C9H13NO3 |
Molecular Weight (g/mol) | 183.21 |
MDL Number | MFCD00008393 |
InChI Key | XEFRNCLPPFDWAC-UHFFFAOYSA-N |
Synonym | benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline |
PubChem CID | 32285 |
IUPAC Name | 3,4,5-trimethoxyaniline |
SMILES | COC1=CC(=CC(=C1OC)OC)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316690
|
Thermo Scientific Acros
139900100 |
10 g | Glass bottle |
£30.90
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10225240
|
Thermo Scientific Acros
139900250 |
25 g | Glass bottle |
£60.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135833
|
Thermo Scientific Acros
139901000 |
100 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24313-88-0 | |
183.21 | |
XEFRNCLPPFDWAC-UHFFFAOYSA-N | |
32285 | |
COC1=CC(=CC(=C1OC)OC)N |
C9H13NO3 | |
MFCD00008393 | |
benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline | |
3,4,5-trimethoxyaniline |
Specifications
24313-88-0 | |
100.0 | |
Pink | |
97% | |
(CH3O)3C6H2NH2 | |
13, 826 | |
Solubility in water: insoluble | |
COC1=CC(=CC(=C1OC)OC)N | |
183.21 | |
183.21 | |
Powder |
96.0 | |
110.0°C to 113.0°C | |
Authentic | |
C9H13NO3 | |
MFCD00008393 | |
benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline | |
XEFRNCLPPFDWAC-UHFFFAOYSA-N | |
3,4,5-trimethoxyaniline | |
32285 | |
97% | |
3, 4, 5-Trimethoxyaniline |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 246-154-4
- 3,4,5-Trimethoxyaniline
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only